missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Axygen™ 0.6mL Self-Standing Screw Cap Tubes
Features O-ring manufactured from specially selected material that is resistant to many commonly-used laboratory chemicals
$2,168.02 - $2,168.02
Specifications
Closure Color | Clear |
---|---|
Closure Material | Polypropylene |
Closure Type | Screw |
Color | Clear |
Material | Polypropylene |
Catalog Number | Mfr. No. | Sterility | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Sterility | Price | Quantity | |||||
14-222-649
|
Corning Incorporated
SCT45SSCS |
Sterile |
Case of 4000 Each for $2,168.02
|
|
Description
For collection, processing, centrifugation, storage and packaging/shipping of restriction enzymes and other reagents.
- Available as presterilized and nonsterile microtubes
- Nonsterile cap tubes have unattached, non-looped caps with O-rings
- Presterilized cap tubes have non-looped caps attached, with O-rings
- Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
- Color: Clear
Specifications
Clear | |
Screw | |
Polypropylene | |
20000xG |
Polypropylene | |
Clear | |
O-ring |