missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Axltide

For cellular and molecular biology applications

Specifications

Product Type Axltide
Molecular Weight 1514.78
Storage Requirements -30° to -10°C
Sequence Lys-Lys-Ser-Arg-Gly-Asp-Tyr-Met-Thr-Met-Gln-Ile-Gly
Format Solid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66170EXD View Documents Alfa Aesar™
J66170EXD
0.5mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ66170MCR View Documents Alfa Aesar™
J66170MCR
1mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Specifications

Axltide
-30° to -10°C
Solid
RUO
1514.78
Lys-Lys-Ser-Arg-Gly-Asp-Tyr-Met-Thr-Met-Gln-Ile-Gly
Axl/Insulin Receptor substrate peptide
HPLC

For Research Use Only