AXL [R499C] Recombinant Human Protein, Invitrogen™ DFS Item
AXL is a transforming gene isolated from primary human myeloid leukemia cells. Recombinant human AXL R499C (473-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Manufacturer:  Life Technologies LSPV6254
 View more versions of this product
Catalog No. PV6254
Description
Description
AXL encodes a novel receptor tyrosine kinase which is a member of the TAM (TYRO3/AXL/MER) receptor tyrosine kinase subfamily. AXL represents a unique structure of the extracellular region that juxtaposes IgL and FNIII repeats and Axl transduces signals from the extracellular matrix into the cytoplasm by binding growth factors like vitamin K-dependent protein growth-arrest-specific gene 6. AXL is involved in the stimulation of cell proliferation and can also mediate cell aggregation by homophilic binding.
  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications
Specifications
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
73.5kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Kinase Assay
Human
GST Tag
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol
100μg
UFO, JTK11
aa 473-end
Yes
Documents