Thermo Scientific™ Automation Tips for Molecular Devices™ Pipetting System
Increase pipetting performance through superior tip straightness and low %CV with these pipette tips for the Molecular Devices system.
Supplier:  Thermo Scientific™ 937262
 View more versions of this product
Catalog No. 21-931-259
Description
Description
Increase pipetting performance with Thermo Scientific™ Automation Tips for Molecular Devices™ Pipetting System, which undergo a rigorous 15-point quality control program to ensure consistency.  Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Molecular Device FLIPR with Type "A" CCS 384-well head and Type "B" Liberty 384-well head.
  • Available in non-sterile, sterile and with the Thermo Scientific™ ART™ filter technology
  • Low percentage coefficient of variation (%CV)
Specifications
Specifications
30 μL
Non-sterile
Rack
Molecular Devices FLIPR Type A CCS 384-well Head
Non-Filtered
Clear
Automation
Molecular Devices
SDS
Documents
Documents
Promotions
Promotions
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.