missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Medegen Autoclavable Waste Bags
These Autoclavable Waste Bags are made of polypropylene.
$171.78 - $305.19
Specifications
Product Type | Autoclavable Bags |
---|---|
Autoclavable | Autoclavable |
Disposable | Yes |
Includes | Indicator, flat pack, twist ties, biohazard labeling, biohazard symbol |
Material | Polypropylene |
Catalog Number | Mfr. No. | Capacity (English) | Dimensions (L x W) | Color | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Dimensions (L x W) | Color | Price | Quantity | |||
14-828-144
|
Medegen
MDG848 |
44 gal. | 38 x 47 in. | Red, Black |
Case of 100 Each for $255.84
|
|
|||
14-828-147
|
Medegen
856 |
20 to 30 gal. | 31 x 38 in. | Red, Black |
Case of 100 Each for $176.09
|
|
|||
14-828-145
|
Medegen
855 |
20 to 30 gal. | 31 x 28 in. | Clear with Red Print |
Case of 100 Each for $171.78
|
|
|||
22-023-800
|
N/A | 12 to 14 gal. | 25 x 30 in. | Red, Black |
Case of 200 Each for $305.19
|
|
|||
Description
- Polypropylene
- 1.8 mil thickness
Specifications
Autoclavable Bags | |
Yes | |
Polypropylene | |
285°F | |
1.80 mil |
Autoclavable | |
Indicator, flat pack, twist ties, biohazard labeling, biohazard symbol | |
0.05 mm | |
140.5°C |