Please login to your online account to display your discounted pricing

Aushon Chemiluminescent Human MMP Array Kits


For simultaneous measurement of matrix metalloproteinase targets

Manufacturer: aushon biosystems inc  11-002-1-FA

 View more versions of this product

Catalog No. 110021FA

  • / Each
 

Description & Specifications

Specifications

Reactivity Human
For Use With (Application) For simultaneous measurement of matrix metalloproteinase targets
Quantity 1 Plex
Product Type MMP 4