Please login to your online account to display your discounted pricing

Aushon Chemiluminescent Human Growth Factor Array Kits


For simultaneous measurement of combinations of growth factors

Manufacturer: aushon biosystems inc  11-010-1-FA

 View more versions of this product

Catalog No. 110101FA

  • / Each
 

Description & Specifications

Specifications

Detects TGFα
For Use With (Application) For simultaneous measurement of combinations of growth factors
Quantity 1 Plex
Product Type Growth Factor 1
Reactivity Human