Please login to your online account to display your discounted pricing

Aushon Chemiluminescent Human Cytokine Array Kits


For simultaneous measurement of combinations of cytokines

Manufacturer: aushon biosystems inc  02-768-1-FA

 View more versions of this product

Catalog No. 027681FA

  • / Each
 

Description & Specifications

Specifications

Quantity 6 Plex
Product Type Cytokine 3
For Use With (Application) For simultaneous measurement of combinations of cytokines
Reactivity Human