Please login to your online account to display your discounted pricing

Aushon Chemiluminescent Human Angiogenesis Array Kits


For simultaneous measurement of combinations of angiogenesis targets

Manufacturer: aushon biosystems inc  09-004-1-FA

 View more versions of this product

Catalog No. 090041FA

  • / Each
 

Description & Specifications

Specifications

Detects PDGF-AA
For Use With (Application) For simultaneous measurement of combinations of angiogenesis targets
Quantity 1 Plex
Product Type Angiogenesis 5
Reactivity Human