Invitrogen™ AURKB (Aurora B) Recombinant Human Protein DFS Item
The Aurora kinases associate with microtubules during chromosome movement and segregation.
Manufacturer:  Invitrogen™ LSPV6131
 View more versions of this product
Catalog No. PV6131
Description
Description
AURKB (Aurora B) localizes to microtubules near kinetochores, specifically to the specialized microtubules called K-fibers.
Specifications
Specifications
NP_004208
Frozen
43.4kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
Aurora Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
AURKB (Aurora B)
Baculovirus
Human
His Tag
Documents