Please login to your online account to display your discounted pricing

Auranofin, Tocris™

Thioredoxin reductase inhibitor; induces MPT

$88.05 - $354.48

Chemical Identifiers

Chemical Name or Material Auranofin
Assay Percent Range >98%
Molecular Formula C20H34AuO9PS
Formula Weight 678.48
Synonym 1-Thio-β-D-glucopyranosatotriethylphosphine gold-2,3,4,6-tetraacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

460050

 
r & d systems
RDS460050
50mg Each for $354.48

460010

 
r & d systems
RDS460010
10mg Each for $88.05
Description & Specifications

Specifications

Formula Weight 678.48
Quantity 50mg
CAS 34031-32-8
Synonym 1-Thio-β-D-glucopyranosatotriethylphosphine gold-2,3,4,6-tetraacetate
Molecular Formula C20H34AuO9PS
Chemical Name or Material Auranofin
Assay Percent Range >98%
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature