Please login to your online account to display your discounted pricing

Attribute Blocks

$41.85 - $41.85

Specifications

Category Mathematics
Format Hands-on Learning
For Use With (Application) Teaches shapes, sorting, patterns, size and counting
Grade PreK+
View More Specs
Products
Catalog Number Mfr. No. Includes Product Type Price Quantity    

S79363B

 
fisher science education
LER1272
Giant set in Plastic storage tray Giant set in Plastic storage tray Each for $41.85
Description & Specifications

Specifications

Category Mathematics
Format Hands-on Learning
For Use With (Application) Teaches shapes, sorting, patterns, size and counting
Grade PreK+

Desk set sizes range from 10cm2 to 30cm2. Giant set sizes range from 25cm2 to 75cm2.