Learn More
Atropine sulfate monohydrate, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1023618
Description
Antagonist at muscarinic receptors; also a bronchodilator. It is also used in used in preanesthetic medication.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAntagonist at muscarinic receptors; also a bronchodilator. It is also used in used in preanesthetic medication.
Solubility
Soluble in water (2500 mg/ml) at 4°C, ethanol (200 mg/ml) at 4°C, Glycerol (400 mg/ml), and methanol.
Notes
Light Sensitive Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Atropine sulfate monohydrate | |
187°C to 192°C | |
MFCD00074815 | |
UN1544 | |
Light sensitive | |
atropine sulfate hydrate, atropine sulfate monohydrate, 1r,3s,5s-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 3-hydroxy-2-phenylpropanoate; methane; sulfuric acid | |
PVGPXGKNDGTPTD-IJTOKZDFSA-N | |
[(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 3-hydroxy-2-phenylpropanoate;sulfuric acid;hydrate | |
23624044 | |
≥98% |
5908-99-6 | |
C17H27NO8S | |
50 g | |
6109275 | |
14,875 | |
Soluble in water (2500mg/ml) at 4°C,ethanol (200mg/ml) at 4°C,Glycerol (400mg/ml),and methanol. | |
CN1C2CCC1CC(C2)OC(=O)C(CO)C3=CC=CC=C3.O.OS(=O)(=O)O | |
405.462 | |
694.84 (676.82 Anhydrous) |
Chemical Identifiers
5908-99-6 | |
405.462 | |
PVGPXGKNDGTPTD-IJTOKZDFSA-N | |
23624044 | |
CN1C2CCC1CC(C2)OC(=O)C(CO)C3=CC=CC=C3.O.OS(=O)(=O)O |
C17H27NO8S | |
MFCD00074815 | |
atropine sulfate hydrate, atropine sulfate monohydrate, 1r,3s,5s-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 3-hydroxy-2-phenylpropanoate; methane; sulfuric acid | |
[(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 3-hydroxy-2-phenylpropanoate;sulfuric acid;hydrate |
Safety and Handling
GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.
P260-P264b-P270-P271-P285-P301+P310-P304+P340-P310-P330-P501c
H300+H330-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.
EINECSNumber : 200-235-0
RTECSNumber : CK2455000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only