missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Atropine sulfate monohydrate, 98+%

Manufacturer:  Alfa Aesar™ A1023609

 View more versions of this product

Catalog No. AAA1023609



Description

Description

Applications
Antagonist at muscarinic receptors; also a bronchodilator. It is also used in used in preanesthetic medication.

Solubility
Soluble in water (2500 mg/ml) at 4°C, ethanol (200 mg/ml) at 4°C, Glycerol (400 mg/ml), and methanol.

Notes
Light Sensitive Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Atropine sulfate monohydrate
10g
C17H27NO8S
UN1544
14,875
PVGPXGKNDGTPTD-IJTOKZDFSA-N
[(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 3-hydroxy-2-phenylpropanoate;sulfuric acid;hydrate
23624044
≥98%
187°C to 192°C
5908-99-6
MFCD00074815
6109275
atropine sulfate hydrate, atropine sulfate monohydrate, 1r,3s,5s-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 3-hydroxy-2-phenylpropanoate; methane; sulfuric acid
CN1C2CCC1CC(C2)OC(=O)C(CO)C3=CC=CC=C3.O.OS(=O)(=O)O
405.462
694.84 (676.82 Anhydrous)

Chemical Identifiers

5908-99-6
405.462
PVGPXGKNDGTPTD-IJTOKZDFSA-N
23624044
CN1C2CCC1CC(C2)OC(=O)C(CO)C3=CC=CC=C3.O.OS(=O)(=O)O
C17H27NO8S
MFCD00074815
atropine sulfate hydrate, atropine sulfate monohydrate, 1r,3s,5s-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 3-hydroxy-2-phenylpropanoate; methane; sulfuric acid
[(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 3-hydroxy-2-phenylpropanoate;sulfuric acid;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.

RTECSNumber : CK2455000

TSCA : TSCA

SDS Documents