Please login to your online account to display your discounted pricing

Alfa Aesar™ Atrial Natriuretic Peptide (11-30), Frog

For cellular and molecular biology applications

Specifications

Product Type Atrial Natriuretic Peptide (11-30), frog
Cross Reactivity Frog
Molecular Weight 2116.40
Storage Requirements -30° to -10°C
Format Lyophilized powder
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66748EXD

 
alfa aesar™
J66748EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66748MCR

 
alfa aesar™
J66748MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Atrial Natriuretic Peptide (11-30), frog
Cross Reactivity Frog
Molecular Weight 2116.40
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence Cys-Phe-Gly-Ser-Arg-Ile-Asp-Arg-Ile-Gly-Ala-Gln-Ser-Gly-Met-Gly-Cys-Gly-Arg-Phe (Disulfide bridge Cys11-Cys27)
Common Name ANP (11-30), frog
Regulatory Status RUO
Purification Method HPLC