Please login to your online account to display your discounted pricing

Alfa Aesar™ Atrial Natriuretic Peptide (1-28), Human

For cellular and molecular biology applications

Specifications

Product Type Atrial Natriuretic Peptide (1-28), human
Cross Reactivity Human
Molecular Weight 3080.47
Storage Requirements -30° to -10°C
Sequence ANP (1-28), human
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66375EXD

 
alfa aesar™
J66375EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66375MCR

 
alfa aesar™
J66375MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Atrial Natriuretic Peptide (1-28), human
Cross Reactivity Human
Molecular Weight 3080.47
Storage Requirements -30° to -10°C
Sequence ANP (1-28), human
Format Lyophilized powder
Regulatory Status RUO
Purification Method HPLC