Please login to your online account to display your discounted pricing

Alfa Aesar™ Atrial Natriuretic Peptide (1-28), Biotin labeled, Human, Porcine

For cellular and molecular biology applications

Specifications

Product Type Atrial Natriuretic Peptide (1-28), biotin labeled, human, porcine
Cross Reactivity Human, Porcine
Molecular Weight 3306.73
Storage Requirements -30° to -10°C
Format Lyophilized powder
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66398LB0

 
alfa aesar™
J66398LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66398MA

 
alfa aesar™
J66398MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66398MCR

 
alfa aesar™
J66398MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Atrial Natriuretic Peptide (1-28), biotin labeled, human, porcine
Cross Reactivity Human, Porcine
Molecular Weight 3306.73
Storage Requirements -30° to -10°C
Format Lyophilized powder
Regulatory Status RUO
Purification Method HPLC