missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Atorvastatin hemicalcium salt, Tocris™

HMG-CoA reductase inhibitor

$142.08 - $598.33

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C33H34FN2O5·+Ca
Formula Weight 577.67
Synonym (R, R)-2-(4-Fluorophenyl)-β, δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoic acid hemicalcium salt
Chemical Name or Material Atorvastatin hemicalcium salt
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
377650 R & D Systems
377650
50mg Each for $598.33
377610 R & D Systems
377610
10mg Each for $142.08
Specifications Description & Specifications

Chemical Identifiers

>99%
577.67
Atorvastatin hemicalcium salt
C33H34FN2O5·+Ca
(R, R)-2-(4-Fluorophenyl)-β, δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoic acid hemicalcium salt

Specifications

134523-03-8
C33H34FN2O5·+Ca
(R, R)-2-(4-Fluorophenyl)-β, δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoic acid hemicalcium salt
Atorvastatin hemicalcium salt
>99%
577.67
50mg

Safety and Handling

Recommended Storage : Store at +4°C