missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
SHOWA™ Triple-Dipped PVC Gloves with Aramid Fiber Lining
Gloves provide protection against a wide range of hazards including cuts, abrasions, chemicals, and oils. Rough textured surface increases durability and enhances grip capabilities.
Supplier: SHOWA™ KV660M08
Description
- Triple dipped PVC coating protects against oils and chemicals
- Kevlar™ aramid fiber liner provides cut resistance
- Rough textured surface enhances grip capabilities
- Hazard barrier is maintained even after multiple washes
- ANSI/ISEA 105-16 Level 4 Puncture Resistance
Specifications
ANSI 6 | |
PVC | |
ANSI A2 | |
Specific | |
Medium | |
JKL | |
Open | |
KV660 | |
30.5 cm |
EN 420:2003 + A1:2009 | |
Blue | |
Fully Textured | |
Dip-coated Gloves | |
4X42B | |
Chemical Spray, Coating Preparation, Drainage, HVAC, Plumbing, and Painting | |
Aramid Fiber | |
12 in. |