Please login to your online account to display your discounted pricing

Atipamezole hydrochloride, Tocris

Selective α2 antagonist

$210.55 - $895.76

Chemical Identifiers

Chemical Name or Material Atipamezole hydrochloride
Assay Percent Range >99%
Molecular Formula C14H16N2·HCl
Formula Weight 248.75
Synonym 5-(2-Ethyl-2, 3-dihydro-1H-indene-2-yl)-1H-imidazole hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

293710

 
r & d systems
RDS293710
10mg Each for $210.55

293750

 
r & d systems
RDS293750
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 248.75
Quantity 10mg
Chemical Name or Material Atipamezole hydrochloride
Assay Percent Range >99%
Molecular Formula C14H16N2·HCl
CAS 104075-48-1
Synonym 5-(2-Ethyl-2, 3-dihydro-1H-indene-2-yl)-1H-imidazole hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature