missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Atenolol, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC449550050
Specifications
Atenolol | |
146.0°C to 148.0°C | |
29122-68-7 | |
C14H22N2O3 | |
atehexal, atenolol, betablok, blokium, cuxanorm, normiten, prenormine, tenoblock, tenormin, tenormine | |
METKIMKYRPQLGS-UHFFFAOYSA-N | |
2-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetamide | |
2249 | |
266.34 |
Glass bottle | |
5g | |
98% | |
15,85 | |
Solubility in water: slightly soluble. Other solubilities: soluble in methanol, acetic acid, dmso, ethanol, insoluble in acetonitrile, ethylacetate, chlorofor, m | |
CC(C)NCC(COC1=CC=C(C=C1)CC(=O)N)O | |
266.34 | |
CHEBI:2904 | |
98% |
Chemical Identifiers
29122-68-7 | |
266.34 | |
atehexal, atenolol, betablok, blokium, cuxanorm, normiten, prenormine, tenoblock, tenormin, tenormine | |
CHEBI:2904 | |
CC(C)NCC(COC1=CC=C(C=C1)CC(=O)N)O |
C14H22N2O3 | |
METKIMKYRPQLGS-UHFFFAOYSA-N | |
2249 | |
2-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetamide |
Safety and Handling
EINECSNumber : 249-451-7