Please login to your online account to display your discounted pricing

ATC 0175 hydrochloride, Tocris

MCH1 antagonist; also 5-HT2B antagonist and 5-HT1A partial agonist

$264.88 - $1,099.17

Chemical Identifiers

Chemical Name or Material ATC 0175 hydrochloride
Assay Percent Range >98%
Molecular Formula C23H25F2N5O·HCl
Formula Weight 461.94
Synonym N-[cis-4-[[4-(Dimethylamino)-2-quinazolinyl]amino]cyclohexyl]-3, 4-difluorobenzamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

334650

 
r & d systems
334650
50mg Each for $1,099.17

334610

 
r & d systems
334610
10mg Each for $264.88
Description & Specifications

Specifications

Formula Weight 461.94
Quantity 50mg
Chemical Name or Material ATC 0175 hydrochloride
Assay Percent Range >98%
Molecular Formula C23H25F2N5O·HCl
CAS 510733-97-8
Synonym N-[cis-4-[[4-(Dimethylamino)-2-quinazolinyl]amino]cyclohexyl]-3, 4-difluorobenzamide hydrochloride
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C