Please login to your online account to display your discounted pricing

ATC 0065, Tocris

Potent and selective MCH1 antagonist

$253.93 - $1,070.58

Chemical Identifiers

Chemical Name or Material ATC 0065
Assay Percent Range >98%
Molecular Formula C25H29BrF3N5O·2HCl
Formula Weight 625.35
Synonym N2-[cis-4-[[2-[4-Bromo-2-(trifluoromethoxy)phenyl]ethyl]amino]cyclohexyl]-N4, N4-dimethyl-2, 4-quinazolinediamine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

335950

 
r & d systems
335950
50mg Each for $1,070.58

335910

 
r & d systems
335910
10mg Each for $253.93
Description & Specifications

Specifications

Formula Weight 625.35
Quantity 50mg
Chemical Name or Material ATC 0065
Assay Percent Range >98%
Molecular Formula C25H29BrF3N5O·2HCl
CAS 510732-84-0
Synonym N2-[cis-4-[[2-[4-Bromo-2-(trifluoromethoxy)phenyl]ethyl]amino]cyclohexyl]-N4, N4-dimethyl-2, 4-quinazolinediamine dihydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature