Please login to your online account to display your discounted pricing

AT 56, Tocris™

L-PGDS inhibitor

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material AT 56
Assay Percent Range >99%
Molecular Formula C25H27N5
Formula Weight 397.52
Synonym 4-(5H-Dibenzo[a, d]cyclohepten-5-ylidene)-1-[4-(2H-tetrazol-5-yl)butyl]-piperidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

353110

 
r & d systems
RDS353110
10mg Each for $205.70

353150

 
r & d systems
RDS353150
50mg Each for $862.95
Description & Specifications

Specifications

Formula Weight 397.52
Quantity 10mg
Chemical Name or Material AT 56
Synonym 4-(5H-Dibenzo[a, d]cyclohepten-5-ylidene)-1-[4-(2H-tetrazol-5-yl)butyl]-piperidine
Assay Percent Range >99%
Molecular Formula C25H27N5
CAS 162640-98-4
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C