Please login to your online account to display your discounted pricing

AT 1015, Tocris™

Long-acting 5-HT2A antagonist

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material AT 1015
Assay Percent Range >97%
Molecular Formula C29H33N3O2·HCl
Formula Weight 492.05
Synonym N-[2-[4-(5H-Dibenzo[a, d]cyclohepten-5-ylidene)-1-piperidinyl]ethyl]-1-formyl-4-piperidinecarboxamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

446810

 
r & d systems
RDS446810
10mg Each for $221.01

446850

 
r & d systems
RDS446850
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 492.05
Quantity 10mg
Chemical Name or Material AT 1015
Assay Percent Range >97%
Molecular Formula C29H33N3O2·HCl
Synonym N-[2-[4-(5H-Dibenzo[a, d]cyclohepten-5-ylidene)-1-piperidinyl]ethyl]-1-formyl-4-piperidinecarboxamide hydrochloride
Recommended Storage Store at +4°C
CAS 190508-50-0
Safety and Handling

Store at +4°C