Please login to your online account to display your discounted pricing

American Educational Products Astrolabe Kit


Perfect for introductory classes in astronomy!

Manufacturer: american educational products  HUB3015

 View more versions of this product

Catalog No. S53032A

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Plotting, Locating movements of stars
Includes Instructions
Quantity 15 kits
Grade 7 to 12
Product Type Astrolabe kit