American Educational Products Astrolabe Kit
Perfect for introductory classes in astronomy!
Manufacturer:  American Educational Products HUB3014
 View more versions of this product
Catalog No. S53032
Description
Description
  • Measures declination, altitude, and azimuth of planets, stars, moon, and locations on the earth
  • Plots and locates movements of stars
  • May also be used as an inclinometer to measure angles of slope, strike, and dip, and for numerous other measurements
Specifications
Specifications
Plotting, Locating movements of stars
Instructions
1 kit
7 to 12
Astrolabe kit
Education
Documents