missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ASTM Type II WATER, Distilled, Deionized, Intermountain

ASTM Type II, Grade B water is prepared by multi-effect distillation designed to produce water having an inline conductivity of 1.0 μS/cm at 25°C.

Manufacturer:  Intermountain Life WRGASTM20L

 View more versions of this product

Catalog No. WRGASTM20L



Description

Description

  • Extremely low bioburden and endotoxin levels
  • Manufactured under cGMP and ISO 13485 guidelines in a FDA registered facility
  • Purified utilizing a nine-step process resulting in the highest quality water on the market
  • Distilled
  • Deionized
Specifications

Specifications

7732-18-5
Clear, Colorless Liquid
H2O
O
18.02
Cubitainer
Yes
GMP and ISO 13485:2016
< 0.25 EU/mL
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
ASTM
Non-Sterile
20 L

Chemical Identifiers

7732-18-5
18.02
water
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
O
Safety and Handling

Safety and Handling

ShelfLife : 36 Months

Recommended Storage : 2-30°C

SDS
Documents