Thermo Scientific™ Assembled Caps and Septa for 8-425 Standard-Opening Vials
Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials.
$22.51 - $105.55
Specifications
Vial Closure Size 8-425mm
For Use With 8-425 Standard-Opening Vials
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Closure Color Closure Material Septum Price Quantity  
Catalog Number Mfr. No. Closure Color Closure Material Septum Price Quantity  
03-375-18D Thermo Scientific™
C40135A
Natural Polypropylene Integral Polypropylene Pack of 100 Each for $22.51
Case of 10 Pack for $225.10
03-378-352 Thermo Scientific™
C401361A
Black Polypropylene Kim-Lok Red PTFE/White Silicone Pack of 100 Each for $98.66
03-378-353 Thermo Scientific™
C401361B
Blue Polypropylene Kim-Lok Red PTFE/White Silicone Pack of 100 Each for $99.11
03-378-355 Thermo Scientific™
C401361G
Green Polypropylene Kim-Lok Red PTFE/White Silicone Pack of 100 Each for $105.55
03-378-354 Thermo Scientific™
C401361R
Red Polypropylene Kim-Lok Red PTFE/White Silicone Pack of 100 Each for $99.57
03-378-357 Thermo Scientific™
C401361W
White Polypropylene Kim-Lok Red PTFE/White Silicone Pack of 100 Each for $99.57
03-378-356 Thermo Scientific™
C401361Y
Yellow Polypropylene Kim-Lok Red PTFE/White Silicone Pack of 100 Each for $99.57
03-378-318 Thermo Scientific™
C401371A
Black Phenolic Ivory PTFE/Red Rubber Pack of 100 Each for $59.02
Specifications
Description
Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials for a wide range of applications.
  • Open top caps can be used with any 8mm septa
  • Phenolic caps are suitable for autoclaving and low temperature applications
  • Polypropylene caps are chemically inert and suitable for most chromatography applications
  • Kim-Lok™ cap/septum assemblies feature a molded centering ring to eliminate septum movement during injection
  • PTFE/red rubber septa are economical for many GC applications
  • PTFE/silicone septa are most popular for HPLC and IC applications
  • Pre-slit septa are easier to pierce with thin-gauge needles
Description & Specifications
Specifications
8-425mm
8-425 Standard-Opening Vials