Please login to your online account to display your discounted pricing
Thermo Scientific™ Assembled Caps and Septa for 8-425 Standard-Opening Vials
Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials.
Manufacturer: thermo scientific™  C401361R
 View more versions of this product
Catalog No. 03-378-354
  • / Pack of 100 Each
Description & Specifications
Specifications
Closure Color Red
Septum Kim-Lok Red PTFE/White Silicone
Type Kim-Lok cap w/flange
For Use With 8-425 Standard-Opening Vials
Item Description C4013-61R
Description PP, Red; Kim-Lok Red PTFE/White Silicone septum
Closure Material Polypropylene
Vial Closure Size 8-425mm
Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials for a wide range of applications.
  • Open top caps can be used with any 8mm septa
  • Phenolic caps are suitable for autoclaving and low temperature applications
  • Polypropylene caps are chemically inert and suitable for most chromatography applications
  • Kim-Lok™ cap/septum assemblies feature a molded centering ring to eliminate septum movement during injection
  • PTFE/red rubber septa are economical for many GC applications
  • PTFE/silicone septa are most popular for HPLC and IC applications
  • Pre-slit septa are easier to pierce with thin-gauge needles