Thermo Scientific™ Assembled Caps and Septa for 8-425 Standard-Opening Vials
Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials.
Manufacturer:  Thermo Scientific™ C401361G
 View more versions of this product
Catalog No. 03-378-355
Description
Description
Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials for a wide range of applications.
  • Open top caps can be used with any 8mm septa
  • Phenolic caps are suitable for autoclaving and low temperature applications
  • Polypropylene caps are chemically inert and suitable for most chromatography applications
  • Kim-Lok™ cap/septum assemblies feature a molded centering ring to eliminate septum movement during injection
  • PTFE/red rubber septa are economical for many GC applications
  • PTFE/silicone septa are most popular for HPLC and IC applications
  • Pre-slit septa are easier to pierce with thin-gauge needles
Specifications
Specifications
Green
8-425mm
8-425 Standard-Opening Vials
Kim-Lok cap w/flange
Polypropylene
PP, Green; Kim-Lok Red PTFE/White Silicone septum
Kim-Lok Red PTFE/White Silicone
PTFE, Silicone
Documents