Please login to your online account to display your discounted pricing
Thermo Scientific™ Assembled Caps and Septa for 8-425 Standard-Opening Vials
Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials.
Manufacturer: thermo scientific™  C401361B
 View more versions of this product
Catalog No. 03-378-353
  • / Pack of 100 Each
Description & Specifications
Specifications
For Use With 8-425 Standard-Opening Vials
Closure Color Blue
Closure Material Polypropylene
Item Description C4013-61B
Vial Closure Size 8-425mm
Description PP, Blue; Kim-Lok Red PTFE/White Silicone septum
Septum Kim-Lok Red PTFE/White Silicone
Type Kim-Lok cap w/flange
Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials for a wide range of applications.
  • Open top caps can be used with any 8mm septa
  • Phenolic caps are suitable for autoclaving and low temperature applications
  • Polypropylene caps are chemically inert and suitable for most chromatography applications
  • Kim-Lok™ cap/septum assemblies feature a molded centering ring to eliminate septum movement during injection
  • PTFE/red rubber septa are economical for many GC applications
  • PTFE/silicone septa are most popular for HPLC and IC applications
  • Pre-slit septa are easier to pierce with thin-gauge needles