Please login to your online account to display your discounted pricing

Alfa Aesar™ Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66909EXD

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66909EXD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 0.5mg
Format Lyophilized powder
Sequence DYKDDDDK Peptide
Regulatory Status RUO
Storage Requirements -30° to -10°C
Common Name Flag PeptideÖ Sigma-Aldrich
Product Type Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys
Molecular Weight 1013.00
Purification Method HPLC