missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ascorbic Acid USP, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Formula Weight 176.13
Chemical Name or Material Ascorbic Acid
Grade USP
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK1852100 Avantor Performance
185257
Glass 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK1852500 Avantor Performance
185210
Glass 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

50-81-7
176.13
USP
C6H8O6
Ascorbic Acid

Specifications

50-81-7
Glass
100g
USP
C6H8O6
176.13
Ascorbic Acid