Please login to your online account to display your discounted pricing

Ascorbic Acid, Reagent Grade, LabChem™

$64.91 - $64.91

Chemical Identifiers

Chemical Name or Material Ascorbic Acid
CAS 50-81-7
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula C6H8O6
Formula Weight 176.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC115309

 
lab chem
LC115309
Amber Glass 100g Each for $64.91
Description & Specifications

Specifications

Formula Weight 176.13
Chemical Name or Material Ascorbic Acid
EINECS Number 200-066-2
Identification Passes Test
Purity Grade Reagent Grade
Physical Form Powder
Packaging Amber Glass
Linear Formula C6H8O6
CAS 50-81-7
Solubility Soluble in water
Density 1.65g/cm3
CAS Max % 100
Health Hazard 2 GHS H Statement
Product is not hazardous.
Melting Point 190°C
Color White
Quantity 100g
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Specific Gravity 1.65g/cm3
Recommended Storage Room Temperature
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Product is not hazardous.

Room Temperature