Please login to your online account to display your discounted pricing

Ascorbic Acid, ACS Reagent Grade, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Ascorbic Acid
CAS 50-81-7
CAS Min % 98.0
CAS Max % 100.0
Purity Grade ACS Reagent Grade
Physical Form Powder or Crystals
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RDCA0750100

 
ricca chemical company
RIRDCA0750100B1
Poly Wide mouth 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder or Crystals
Purity Grade ACS Reagent Grade
Color White or white
Quantity 100g
CAS 50-81-7
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material Ascorbic Acid
Packaging Poly Wide mouth