missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ascarite(II), CO2 absorbent, 20-30 mesh, capacity: 40-50%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC208091000

 View more versions of this product

Catalog No. AC208091000


 

Specifications Description & Specifications

Specifications

Ascarite(II)™, CO2 absorbent, 20-30 mesh, capacity: 40-50%
100g
81133-20-2,1310-73-2,7631-86-9
Particle size analysis: 20 to 30 mesh
MFCD00132264
Beige to light brown
20- to 30-mesh
Glass bottle
Granules

Chemical Identifiers

81133-20-2,1310-73-2,7631-86-9
MFCD00132264
Ascarite(II)™, CO2 absorbent, 20-30 mesh, capacity: 40-50%, 20- to 30-mesh

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger