Please login to your online account to display your discounted pricing

Tocris™ AS 605240

Potent and selective PI 3-kinase gamma (PI 3-Kgamma) inhibitor

$195.49 - $816.90

Specifications

Product Type AS 605240
Inhibitors AS 605240
Molecular Weight 257.27
Purity >98
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

357810R

 
tocris™
RDS357810
10mg Each for $195.49

357850R

 
tocris™
RDS357850
50mg Each for $816.90
Description & Specifications

Specifications

Product Type AS 605240
Inhibitors AS 605240
Molecular Weight 257.27
Purity >98

Potent and selective inhibitor of PI 3-kinase γ (PI 3-Kγ) (IC50 = 8nM). Displays 30-fold selectivity over PI 3-Kδ and PI 3-Kβ and 7.5-fold selectivity over PI 3-Kα. Suppresses the progression of joint inflammation and damage in both lymphocyte-independent and lymphocyte-dependent mouse models of rheumatoid arthritis. Orally active and ATP-competitive.

  • Chemical Name: 5-(6-Quinoxalinylmethylene)-2,4-thiazolidine-2,4-dione
  • Formula: C12H7N3O2S
  • CAS Number: 648450-29-7
  • Solubility: Soluble to 1mM in DMSO with gentle warming
  • Purity: >98