Please login to your online account to display your discounted pricing

AS 19, Tocris

Reported potent 5-HT7 agonist

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material AS 19
Assay Percent Range >98%
Molecular Formula C18H25N3
Formula Weight 283.41
Synonym (2S)-(+)-5-(1, 3, 5-Trimethylpyrazol-4-yl)-2-(dimethylamino)tetralin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

196810

 
r & d systems
RDS196810
10mg Each for $244.81

196850

 
r & d systems
RDS196850
50mg Each for $997.23
Description & Specifications

Specifications

Formula Weight 283.41
Quantity 10mg
Chemical Name or Material AS 19
Assay Percent Range >98%
Molecular Formula C18H25N3
CAS 1000578-26-6
Synonym (2S)-(+)-5-(1, 3, 5-Trimethylpyrazol-4-yl)-2-(dimethylamino)tetralin
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C