Please login to your online account to display your discounted pricing

AS 1892802, Tocris™

Potent ROCK inhibitor; orally bioavailable

$330.06 - $1,362.21

Chemical Identifiers

Chemical Name or Material AS 1892802
Assay Percent Range >99%
Molecular Formula C20H19N3O2
Formula Weight 333.38
Synonym N-[(1S)-2-Hydroxy-1-phenylethyl]-N'-[4-(4-pyridinyl)phenyl]-urea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

492750

 
r & d systems
RDS492750
50mg Each for $1,362.21

492710

 
r & d systems
RDS492710
10mg Each for $330.06
Description & Specifications

Specifications

Formula Weight 333.38
Quantity 50mg
Chemical Name or Material AS 1892802
Assay Percent Range >99%
Molecular Formula C20H19N3O2
CAS 928320-12-1
Synonym N-[(1S)-2-Hydroxy-1-phenylethyl]-N'-[4-(4-pyridinyl)phenyl]-urea
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature