Please login to your online account to display your discounted pricing

AS 1269574, Tocris™

GPR119 receptor agonist

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material AS 1269574
Assay Percent Range >98%
Molecular Formula C13H14BrN3O
Formula Weight 308.17
Synonym 2-[[2-(4-Bromophenyl)-6-methyl-4-pyrimidinyl]amino]ethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

417710

 
r & d systems
417710
10mg Each for $225.77

417750

 
r & d systems
417750
50mg Each for $924.23
Description & Specifications

Specifications

Formula Weight 308.17
Quantity 10mg
Chemical Name or Material AS 1269574
Assay Percent Range >98%
Molecular Formula C13H14BrN3O
CAS 330981-72-1
Synonym 2-[[2-(4-Bromophenyl)-6-methyl-4-pyrimidinyl]amino]ethanol
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C