missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

AS 101, Tocris

Immunomodulator; inhibits IL-10 synthesis and potentiates IL-1α, IL-2 and TNF-α release

$168.15 - $697.40

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C2H4Cl3O2Te·NH4
Formula Weight 312.05
Synonym Ammonium trichloro[1, 2-ethanediolato-O, O']-tellurate
Chemical Name or Material AS 101
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
244650 R & D Systems
RDS244650
50mg Each for $697.40
244610 R & D Systems
RDS244610
10mg Each for $168.15
Specifications Description & Specifications

Chemical Identifiers

>98%
312.05
AS 101
C2H4Cl3O2Te·NH4
Ammonium trichloro[1, 2-ethanediolato-O, O']-tellurate

Specifications

106566-58-9
C2H4Cl3O2Te·NH4
Ammonium trichloro[1, 2-ethanediolato-O, O']-tellurate
AS 101
>98%
312.05
50mg

Safety and Handling

Recommended Storage : Desiccate at Room Temperature