Please login to your online account to display your discounted pricing

Arvanil, Tocris


Potent CB1 and TRPV1 agonist; Also anandamide transport inhibitor

Quantity 25mg

Manufacturer: r & d systems  RDS135425

 View more versions of this product

Catalog No. 135425

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 439.64
Quantity 25mg
Assay Percent Range >98%
Molecular Formula C28H41NO3
Synonym N-[(4-Hydroxy-3-methoxyphenyl)methyl]-5Z,8Z,11Z,14Z-eicosatetraenamide
Chemical Name or Material Arvanil
CAS 128007-31-8
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material Arvanil
Assay Percent Range >98%
Molecular Formula C28H41NO3
Formula Weight 439.64
Synonym N-[(4-Hydroxy-3-methoxyphenyl)methyl]-5Z,8Z,11Z,14Z-eicosatetraenamide
Accessories

Desiccate at -20°C