Thermo Scientific™ ART™ Barrier Reload System PROMO
Perform applications accurately and consistently without compromising protection with this barrier reload system.
$1,695.62 - $1,695.62
Specifications
Clean Claims DNase, RNase, ATP, and Pyrogen Free
Color Crystal
Sterility Sterile
Model ART Barrier
View More Specs
Products 1
Catalog Number Mfr. No. Volume (Metric) Description Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Description Price Quantity  
13-811-100PR View Documents Thermo Scientific™
PR2139RT
10μL ART 10 Barrier Tip Case of 5 Each for $1,695.62
Description
Description
  • ART barrier tips provide 100% security against aerosols and liquid contamination
  • Stackable barrier tip packaging allows for tips to pass through offset holes in lower insert
  • Blister sleeve guide rails align tip inserts for smooth transfer into the rack
  • Outer sleeve protects tips from exterior contamination while refilling inserts
  • Tip packaging option reduces plastic waste
  • Rigid insert design provides reliable tip loading
  • Lot number and description are printed on every insert
  • Specifications
    Specifications
    DNase, RNase, ATP, and Pyrogen Free
    Crystal
    Sterile
    ART Barrier