missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Arsenious Acid Solution, For Iodide, LabChem™

$54.46 - $54.46

Chemical Identifiers

CAS 7732-18-5,1327-53-3,7664-93-9
CAS Max % 99.46,0.5,0.04
Linear Formula As2O3
Formula Weight 197.84
Chemical Name or Material Arsenious Acid Solution, For Iodide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC115201 Lab Chem
LC115201
500mL Each for $54.46
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,1327-53-3,7664-93-9
As2O3
Arsenious Acid Solution, For Iodide
99.46,0.5,0.04
197.84

Specifications

7732-18-5,1327-53-3,7664-93-9
Liquid
197.84
Passes Test
Soluble in water
Room Temperature
Clear, colorless
Arsenious Acid Solution, For Iodide
99.46,0.5,0.04
As2O3
1g/mL
Poly Bottle
1g/mL
Arsenic and its oxides; Sulfur compounds
500mL

Safety and Handling

GHS H Statement
May cause cancer.

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe dust.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger