Please login to your online account to display your discounted pricing

Arsenic(V) oxide, 99.9+%, (trace metal basis), ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC366310050

 View more versions of this product

Catalog No. AC366310050

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Trace Metal Basis
Physical Form Powder and Chunks
Formula Weight 229.84
Quantity 5g
Color White to off-white
Molecular Formula As2O5
Total Trace Metal Impurities 0.1% max.
CAS 1303-28-2
Linear Formula As2O5
Synonym Arsenic pentoxide
Packaging Glass bottle
Health Hazard 1 Danger
Chemical Name or Material Arsenic(V) oxide
Name Note 99.9+%
Assay Percent Range 99.9+%
MDL Number MFCD00003434
Health Hazard 2 GHS H Statement:
Toxic if inhaled.
May cause cancer.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage Poison Room

Specifications

Chemical Name or Material Arsenic(V) oxide
Name Note 99.9+%
CAS 1303-28-2
Purity Grade Trace Metal Basis
Assay Percent Range 99.9+%
Molecular Formula As2O5
Linear Formula As2O5
Formula Weight 229.84
MDL Number MFCD00003434
Synonym Arsenic pentoxide
Accessories

Danger

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if inhaled.
May cause cancer.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.

Poison Room

SDS & Safety Documents