Please login to your online account to display your discounted pricing

Arsenic(V) oxide, 99.9+%, (trace metal basis), ACROS Organics™

$70.10 - $70.10

Chemical Identifiers

Chemical Name or Material Arsenic(V) oxide
Name Note 99.9+%
CAS 1303-28-2
Purity Grade Trace Metal Basis
Assay Percent Range 99.9+%
Molecular Formula As2O5
Linear Formula As2O5
Formula Weight 229.84
MDL Number MFCD00003434
Synonym Arsenic pentoxide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC366310050

  SDS
thermo fisher scientific
AC366310050
Glass bottle 5g Each for $70.10
Description & Specifications

Specifications

Purity Grade Trace Metal Basis
Formula Weight 229.84
Physical Form Powder and Chunks
Quantity 5g
Color White to off-white
Total Trace Metal Impurities 0.1% max.
Molecular Formula As2O5
CAS 1303-28-2
Synonym Arsenic pentoxide
Linear Formula As2O5
Packaging Glass bottle
Health Hazard 1 Danger
Chemical Name or Material Arsenic(V) oxide
Assay Percent Range 99.9+%
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00003434
Name Note 99.9+%
Health Hazard 2 GHS H Statement:
Toxic if inhaled.
May cause cancer.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Recommended Storage Poison Room

For ultra trace analysis

Safety and Handling

Danger

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if inhaled.
May cause cancer.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.

Poison Room