Please login to your online account to display your discounted pricing

Arsenic Trioxide, Reagent Grade, ≥ 99.0%, LabChem™

$199.40 - $199.40

Chemical Identifiers

Chemical Name or Material Arsenic Trioxide
CAS 1327-53-3
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula As2O3
Formula Weight 197.84
UN Number UN1561
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC115059

 
lab chem
LC115059
100g Each for $199.40
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 197.84
Physical Form Powder
Decomposition Information Arsenic and its oxides
Health Hazard 1 Danger
Linear Formula As2O3
CAS Max % 100
CAS 1327-53-3
UN Number UN1561
Quantity 100g
Packaging Amber Glass
Boiling Point 102°C
Melting Point −7°C
Color White
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe dust.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Identification Passes Test
Solubility Soluble in water
EINECS Number 215-481-4
Health Hazard 2 GHS H Statement
Fatal if swallowed.
Causes severe skin burns and eye damage.
May cause cancer.
Toxic to aquatic life.
Storage Information Room Temperature
Specific Gravity 1.02g/mL
Chemical Name or Material Arsenic Trioxide
Purity Grade Reagent Grade
Density 1.02g/mL
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe dust.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.

GHS H Statement
Fatal if swallowed.
Causes severe skin burns and eye damage.
May cause cancer.
Toxic to aquatic life.