missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Arsenic standard solution, for AAS, 1mg/mL As in 2% KOH, ACROS Organics™

Manufacturer:  Acros Organics 195821000

 View more versions of this product

Catalog No. AC195821000



Description

Description

for atomic adsorption spectroscopy
Specifications

Specifications

Arsenic standard solution
7440-38-2,1310-58-3,7732-18-5
MFCD00085309
15, 784
[As]
74.922
CHEBI:27563
Solution
Plastic bottle
Undesignated
1 mg/ml As in 2% KOH
As
997 to 1003 μg/mL of As(3+) in 2% KOH (20°C)
RQNWIZPPADIBDY-UHFFFAOYSA-N
arsenic
5359596
74.92
AAS
1.02g/mL
100mL

Chemical Identifiers

7440-38-2
74.922
RQNWIZPPADIBDY-UHFFFAOYSA-N
CHEBI:27563
[As]
As
MFCD00085309
5359596
arsenic
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents