Please login to your online account to display your discounted pricing

Alfa Aesar™ Arsenic sponge, 30mm (1.18 in.) & down, 99.5% (metals basis)

Chemical Identifiers

Chemical Name or Material Arsenic sponge
CAS 7440-38-2
MDL Number MFCD00085309
UN Number UN1558
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3973214

  SDS
alfa aesar™
3973214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3973222

  SDS
alfa aesar™
3973222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Irregular Pieces
Quantity 25g
Health Hazard 3 GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Arsenic sponge
Odor Odorless
UN Number UN1558
CAS 7440-38-2
Health Hazard 1 Danger
Assay Percent Notes (metals basis)
Assay 99.5%
TSCA TSCA
Health Hazard 2 GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
MDL Number MFCD00085309
Particle Size 30mm (1.18 in.) & down
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.